Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Cyclase 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535382 Supplier Invitrogen™ Supplier No. PA535382
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody contains enough material to conduct 10 mini-Western blots.

This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 3
Applications Immunohistochemistry, Western Blot, Immunocytochemistry, Multiplex Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene Adcy3
Gene Accession No. O60266, P21932, Q8VHH7
Gene Alias AC3; AC-III; ADCY3; adenylate cyclase 3; adenylate cyclase 8 (brain); adenylate cyclase type 3; Adenylate cyclase type III; adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; KIAA0511; mKIAA0511; type III adenylyl cyclase
Gene Symbols Adcy3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal region of rat ADCY3 conjugated to KLH.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104111, 109, 64508
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.