Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 3 Polyclonal Antibody
Chicken Polyclonal Antibody
$514.50 - $1763.00
Specifications
Antigen | Adenylate Cyclase 3 |
---|---|
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143525
|
Invitrogen™
PA5143525 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143559
|
Invitrogen™
PA5143559 |
100 μL |
Each for $514.50
|
|
|||||
Description
Adenylate Cyclase 3 Polyclonal Antibody for Western Blot, ICC/IF
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes.Specifications
Adenylate Cyclase 3 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgY | |
Mouse, Rat | |
Adcy3 | |
AC3; AC-III; ADCY3; adenylate cyclase 3; adenylate cyclase 8 (brain); adenylate cyclase type 3; Adenylate cyclase type III; adenylate cyclase, olfactive type; Adenylyl cyclase 3; adenylyl cyclase, type III; ATP pyrophosphate-lyase 3; KIAA0511; mKIAA0511; type III adenylyl cyclase | |
Adcy3 | |
Primary | |
Affinity chromatography |
1 mg/mL | |
Western Blot, Immunocytochemistry, Western Blot | |
Unconjugated | |
Chicken | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
P21932, Q8VHH7 | |
104111, 64508 | |
C-terminal peptide of rat ACIII, PAAFPNGSSVTLPHQVVDNP with a Cys added to the N-terminus to allow coupling to KLH. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title