Learn More
Invitrogen™ Adenylate Cyclase 4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total ADCY4.
Specifications
Specifications
| Antigen | Adenylate Cyclase 4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Adcy4 |
| Gene Accession No. | P26770, Q8NFM4, Q91WF3 |
| Gene Alias | A cyclase IV; AC4; Adcy4; adenylate cyclase 4; adenylate cyclase type 4; Adenylate cyclase type IV; adenylyl cyclase 4; ATP pyrophosphate-lyase 4; KIAA4004; mKIAA4004 |
| Gene Symbols | Adcy4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.