Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561717
Description
Immunogen sequence: FAHLSHGDSP VSTSTPLPEK TLASFSTQWS LDRSRTPRGL DDELDTGDAK FFQVIEQLNS QKQWKQSKDF NPLTLYFREK EMEKEYRLSA IP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.
This gene encodes a member of the family of adenylate cyclases, which are membrane-associated enzymes that catalyze the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). Mouse studies show that adenylate cyclase 4, along with adenylate cyclases 2 and 3, is expressed in olfactory cilia, suggesting that several different adenylate cyclases may couple to olfactory receptors and that there may be multiple receptor-mediated mechanisms for the generation of cAMP signals.
Specifications
Adenylate Cyclase 4 | |
Polyclonal | |
Unconjugated | |
Adcy4 | |
A cyclase IV; AC4; Adcy4; adenylate cyclase 4; adenylate cyclase type 4; Adenylate cyclase type IV; adenylyl cyclase 4; ATP pyrophosphate-lyase 4; KIAA4004; mKIAA4004 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
196883 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NFM4 | |
Adcy4 | |
Recombinant protein corresponding to Human Adenylate Cyclase 4. Recombinant protein control fragment (Product #RP-103021). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction