Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Cyclase 5 Mouse anti-Mouse, Rat, Porcine, Clone: 19D5.C1, MilliporeSigma™

Catalog No. MABS2049100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS2049-100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS2049-100 Supplier MilliporeSigma Supplier No. MABS2049100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Adenylate Cyclase 5, clone 19D5.C1 is a mouse monoclonal antibody that detects adenylate cyclase 5 and has been tested for use in ELISA and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase 5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 19D5.C1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Adcy5
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 20 amino acids from the cytoplasmic domain of murine adenylate cyclase V.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001012783.3
Test Specificity Clone 19D5.C1 detects Adnylate Cyclase 5 in murine, rat, and procine cells. It targets an epitope within 20 amino acids from the cytoplasmic domain.
Target Species Mouse, Rat, Pig
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.