Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567466
Description
Immunogen sequence: FTCNSRDLLGC LAQEHNISAS QVNACHVAES AVNYSLGDEQ GFCGSPWPNC NF Highest antigen sequence identity to the following orthologs - mouse 79%, rat 77%.
This gene encodes a member of the membrane-bound adenylyl cyclase enzymes. Adenylyl cyclases mediate G protein-coupled receptor signaling through the synthesis of the second messenger cAMP. Activity of the encoded protein is stimulated by the Gs alpha subunit of G protein-coupled receptors and is inhibited by protein kinase A, calcium and Gi alpha subunits. Single nucleotide polymorphisms in this gene may be associated with low birth weight and type 2 diabetes. Alternatively spliced transcript variants that encode different isoforms have been observed for this gene. [provided by RefSeq, Dec 2010].
Specifications
Adenylate Cyclase 5 | |
Polyclonal | |
Unconjugated | |
ADCY5 | |
AC5; Adcy5; adenylate cyclase 5; adenylate cyclase type 5; Adenylate cyclase type V; adenylyl cyclase 5; adenylyl cyclase type V; ATP pyrophosphate-lyase 5; AW121902; FDFM | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
111 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95622 | |
ADCY5 | |
Recombinant Human Adenylate Cyclase 5. Recombinant protein control fragment (Product #RP-108121). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction