Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylate Cyclase 7 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297909R
Description
Adenylate Cyclase 7 Polyclonal specifically detects Adenylate Cyclase 7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Adenylate Cyclase 7 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
AC7, adenylate cyclase 7, adenylate cyclase type 7, Adenylate cyclase type VII, Adenylyl cyclase 7, ATP pyrophosphate-lyase 7, EC 4.6.1, EC 4.6.1.1, KIAA0037FLJ36387 | |
Produced in rabbits immunized with E. coli-derived Human Adenylate Cyclase 7 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
113 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction