Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenylate Cyclase 9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536919
Description
This antibody detects endogenous protein at a molecular weight of 161 kDa. Purity is >95% by SDS-PAGE.
Adenylate cyclase is a membrane bound enzyme that catalyses the formation of cyclic AMP from ATP. It is regulated by a family of G protein-coupled receptors, protein kinases, and calcium. The type 9 adenylyl cyclase is a widely distributed adenylyl cyclase, and it is stimulated by beta-adrenergic receptor activation but is insensitive to forskolin, calcium, and somatostatin.
Specifications
Adenylate Cyclase 9 | |
Polyclonal | |
Unconjugated | |
ADCY9 | |
AC9; ACIX; ACtp10; Adcy9; adenylate cyclase 9; adenylate cyclase type 9; Adenylate cyclase type IX; adenylyl cyclase 9; Adenylyl cyclase type 10; ATP pyrophosphate-lyase 9; AW125421; D16Wsu65e; KIAA0520; mKIAA0520; type IX ATP pyrophosphate-lyase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115, 11515, 302950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O60503, P51830 | |
ADCY9 | |
A synthetic peptide corresponding to residues in human ADCY9. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction