Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylate Cyclase Toxin Mouse anti-Bacteria, Clone: 3D1, MilliporeSigma™
Mouse Monoclonal Antibody
$192.35 - $456.56
Specifications
Antigen | Adenylate Cyclase Toxin |
---|---|
Clone | 3D1 |
Concentration | Please refer to lot specific datasheet. |
Applications | ELISA, Flow Cytometry, Inhibition Assays, Neutralization, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2086-25
|
MilliporeSigma
MABF208625UG |
25 μg |
Each for $192.35
|
|
|||||
MABF2086-100
|
MilliporeSigma
MABF2086100UG |
100 μg |
Each for $456.56
|
|
|||||
Description
Anti-Adenylate Cyclase Toxin, B. pertussis, clone 3D1 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis and has been tested for use in ELISA, Flow Cytometry, Inhibition Assay, Neutralizing, and Western Blotting.Specifications
Adenylate Cyclase Toxin | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Bacteria | |
IgG1 κ | |
Protein G Purified | |
Clone 3D1 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis. |
3D1 | |
ELISA, Flow Cytometry, Inhibition Assays, Neutralization, Western Blot | |
Unconjugated | |
Mouse | |
Inflammation and Immunology | |
Adenylate cyclase (AC) toxin from Bordetella pertussis. | |
Primary | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title