Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adenylate Cyclase Toxin Mouse anti-Bacteria, Clone: 9D4, MilliporeSigma™

Catalog No. MABF2087100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2087-100 100 μL
MABF2087-25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF2087-100 Supplier MilliporeSigma Supplier No. MABF2087100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Adenylate Cyclase Toxin, B. pertussis, clone 9D4 Function Assay and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Cyclase Toxin
Applications Dot Blot, Functional Assay, Western Blot
Classification Monoclonal
Clone 9D4
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Host Species Mouse
Immunogen Adenylate cyclase toxin from Bordetella pertussis.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 9D4 is a mouse monoclonal antibody that detects Adenylate cyclase (AC) toxin from Bordetella pertussis.
Target Species Bacteria
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.