Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylate Kinase 1 Antibody (OTI4A1), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270123MFV610
Description
Adenylate Kinase 1 Monoclonal antibody specifically detects Adenylate Kinase 1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow CytometrySpecifications
Adenylate Kinase 1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Monkey | |
Purified |
Flow Cytometry | |
OTI4A1 | |
Flow Cytometry | |
adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase | |
Full length human recombinant protein of human AK1 (NP_000467) produced in HEK293T cell. | |
0.1 mL | |
metabolism, Signal Transduction | |
203 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction