Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylate Kinase 1 Antibody (OTI4A1), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270123PCP
Description
Adenylate Kinase 1 Monoclonal specifically detects Adenylate Kinase 1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
| Adenylate Kinase 1 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| AK1 | |
| Full length recombinant protein of human Adenylate Kinase 1 (NP_000467) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Flow Cytometry | |
| OTI4A1 | |
| Flow Cytometry | |
| adenylate kinase 1, adenylate kinase isoenzyme 1, AK 1, ATP-AMP transphosphorylase 1, EC 2.7.4, EC 2.7.4.3, Myokinase | |
| Mouse | |
| Affinity Purified | |
| metabolism, Signal Transduction | |
| 203.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction