Learn More
Invitrogen™ Adenylate Kinase 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578748
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell lysates, human SiHa whole cell lysates, rat heart tissue lysates, mouse lung tissue lysates, mouse heart tissue lysates. IHC: human colon cancer tissue, human lung adenocarcinoma tissue, mouse heart tissue, rat heart tissue. Flow: PC-3 cell.
Adenylate kinase is an enzyme involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate group among adinine nucleotides. Three isozymes of adenylate kinase have been identified in vertebrates, adenylate isozyme 1 (AK1), 2 (AK2) and 3 (AK3). AK1 is found in the cytosol of skeletal muscle, brain and erythrocytes, whereas AK2 and AK3 are found in the mitochondria of other tissues including liver and heart. AK1 was identified because of its association with a rare genetic disorder causing nonspherocytic hemolytic anemia where a mutation in the AK1 gene was found to reduce the catalytic activity of the enzyme.
Specifications
Adenylate Kinase 1 | |
Polyclonal | |
Unconjugated | |
AK1 | |
adenylate kinase 1; adenylate kinase isoenzyme 1; adenylate kinase isoenzyme 1 {ECO:0000255; Adenylate monophosphate kinase; adenylate monophosphate kinase {ECO:0000255; AK 1; AK 1 {ECO:0000255; Ak1; Ak-1; ATP:AMP phosphotransferase; ATP:AMP phosphotransferase {ECO:0000255; ATP-AMP transphosphorylase 1; ATP-AMP transphosphorylase 1 {ECO:0000255; B430205N08Rik; cytosolic adenylate kinase; HAMAP-Rule:MF_03171}; HTL-S-58j; I79_014777; Myokinase; myokinase {ECO:0000255; testis secretory sperm binding protein Li 58j | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11636, 203, 24183 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P00568, P39069, Q9R0Y5 | |
AK1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human AK1 (149-189aa RLETYYKATEPVIAFYEKRGIVRKVNAEGSVDSVFSQVCTH). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.