Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Kinase 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553813 Supplier Invitrogen™ Supplier No. PA553813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAENFCVCHL ATGDMLRAMV ASGSELGKKL KATMDAGKLV SDEMVVELIE KNLETPLCKN GFLLDGFPRT VRQAEMLDDL MEKRKEKLDS VIEFSIPDSL LIRR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 96%.

Adenylate kinases are involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. Three isozymes of adenylate kinase, namely 1, 2, and 3, have been identified in vertebrates; this gene encodes isozyme 2. Expression of these isozymes is tissue-specific and developmentally regulated. Isozyme 2 is localized in the mitochondrial intermembrane space and may play a role in apoptosis. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AK2
Gene Accession No. P29410, P54819, Q9WTP6
Gene Alias adenylate kinase 2; adenylate kinase 2, mitochondrial; adenylate kinase 2, mitochondrial {ECO:0000255; Adenylate kinase 2, mitochondrial, N-terminally processed; adenylate kinase isoenzyme 2, mitochondrial; adenylate monophosphate kinase; adenylate monophosphate kinase {ECO:0000255; ADK2; AK 2; AK 2 {ECO:0000255; Ak2; Ak-2; ATP:AMP phosphotransferase; ATP:AMP phosphotransferase {ECO:0000255; ATP-AMP transphosphorylase 2; ATP-AMP transphosphorylase 2 {ECO:0000255; D4Ertd220e; HAMAP-Rule:MF_03168}; si:ch211-197n10.1; testis secretory sperm-binding protein Li 220n; wu:fb34f05; wu:fj80e03
Gene Symbols AK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Adenylate Kinase 2. Recombinant protein control fragment (Product #RP-91714).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11637, 204, 24184
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.