Learn More
Invitrogen™ Adenylate Kinase 3 Monoclonal Antibody (OTI4C4)

Description
Adenylate Kinase 3 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | Adenylate Kinase 3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4C4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | AK3 |
| Gene Accession No. | Q9UIJ7 |
| Gene Alias | AA407498; adenylate kinase 3; adenylate kinase 3 {ECO:0000255; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255; adenylate kinase 6, adenylate kinase 3 like 1; AI506714; AK 3; AK 3 {ECO:0000255; AK3; AK-3; Ak3l; AK3L1; AK4; AK6; AKL3L; AKL3L1; FIX; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial; HAMAP-Rule:MF_03169} |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.