Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Adenylate Kinase 4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528578
Description
Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Mouse (88%), Rat (87%), Pig (90%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.
Specifications
Adenylate Kinase 4 | |
Polyclonal | |
Unconjugated | |
AK4 | |
adenylate kinase 3 alpha-like 1; Adenylate kinase 3-like; adenylate kinase 3-like {ECO:0000255; adenylate kinase 3-like 1; adenylate kinase 3-like 2; adenylate kinase 4; adenylate kinase 4, mitochondrial; adenylate kinase 4, mitochondrial {ECO:0000255; Adenylate kinase isoenzyme 4; adenylate kinase isoenzyme 4, mitochondrial; adenylate kinase isoenzyme 4, mitochondrial; adenylate kinase 4, mitochondrial; AK 4; AK 4 {ECO:0000255; AK3; Ak-3; Ak3b; Ak3l1; AK3L2; AK4; Ak-4; ATP-AMP transphosphorylase; D4Ertd274e; GTP:AMP phosphotransferase AK4; GTP:AMP phosphotransferase AK4 {ECO:0000255; GTP:AMP phosphotransferase AK4, mitochondrial; HAMAP-Rule:MF_03170}; mitochondrial adenylate kinase-3; nucleoside-triphosphate-adenylate kinase; RP4-686B20.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11639, 205, 29223 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P27144, Q9WUR9, Q9WUS0 | |
AK4 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 197 of Human Adenylate kinase 3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction