Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Kinase 5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553933
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553933 Supplier Invitrogen™ Supplier No. PA553933
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLFPNKEAAA GSSDLDPSMI LDTGEIIDTG SDYEDQGDDQ LNVFGEDTMG GFMEDLRKCK IIFIIGGPGS GKGTQCEKLV E Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 83%.

This gene encodes a member of the adenylate kinase family, which is involved in regulating the adenine nucleotide composition within a cell by catalyzing the reversible transfer of phosphate groups among adenine nucleotides. This member is related to the UMP/CMP kinase of several species. It is located in the cytosol and expressed exclusively in brain. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AK5
Gene Accession No. Q9Y6K8
Gene Alias adenylate kinase 5; adenylate kinase 6; adenylate kinase isoenzyme 5; AK 5; AK5; AK6; ATP-AMP transphosphorylase 5; MGC33326; wu:fj63a06
Gene Symbols AK5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Adenylate Kinase 5. Recombinant protein control fragment (Product #RP-92479).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26289
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.