Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Kinase 7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109375 Supplier Invitrogen™ Supplier No. PA5109375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAEEEETAAL TEKVIRTQRV FINLLDSYSS GNIGKFLSNC VVGASLEEIT EEEEEEDENK SAMLEASSTK VKEGTFQIVG TLSKPDSPRP DFAVETYSAI SREDLLMRLL ECDVIIYNIT ESSQQMEEAI WAVSALSEEV SHFEKRKLFI LLSTVMTWAR SKALDPEDSE VPFTEEDYRR RKSHPNFLDH INAEKMVLKF.

The adenylate kinases (AK) are a family of structurally and functionally related enzymes that catalyze a similar reaction, MgNTP + AMP = MgNDP + ADP (N = A or G). The AK enzymes are important for maintenance of homeostasis of the adenine and guanine nucleotide pools. AK1 is a cytosolic enzyme for which ATP is the substrate. AK2 catalyzes the same reaction as AK1, but it is localized in the mitochondrial intermembrane space. AK3 is present in the mitochondrial matrix and prefers GTP over ATP as the substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 7
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene AK7
Gene Accession No. Q96M32, Q9D2H2
Gene Alias 4930502N02Rik; adenylate kinase 7; AK 7; AK7; ATP-AMP transphosphorylase 7; putative adenylate kinase 7
Gene Symbols AK7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AK7 (NP_689540.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122481, 314416, 78801
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.