Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Kinase 8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554409
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554409 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554409 Supplier Invitrogen™ Supplier No. PA554409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGERYHLMYK PPPTMEIQAR LLQNPKDAEE QVKLKMDLFY RNSADLEQLY GSAITLNGDQ DPYTVFEYIE SGIINPLPKK IP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

Enables AMP binding activity and nucleobase-containing compound kinase activity. Involved in nucleoside diphosphate phosphorylation and nucleoside triphosphate biosynthetic process. Located in 9+2 motile cilium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 8
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ak8
Gene Accession No. Q96MA6
Gene Alias 1190002A17Rik; Adenylate kinase 8; Adenylate kinase isoenzyme 4, mitochondrial (ATP-AMP transphosphorylase); AK 8; AK8; ATP-AMP transphosphorylase 8; C9orf98; DDX31; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98 homolog; RGD1303144; RP11-143F18.1
Gene Symbols Ak8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Adenylate Kinase 8. Recombinant protein control fragment (Product #RP-93049).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158067
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.