Learn More
Invitrogen™ Adenylate Kinase 8 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Bovine (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Adenylate Kinase 8 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.62 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | Ak8 |
| Gene Accession No. | Q96MA6 |
| Gene Alias | 1190002A17Rik; Adenylate kinase 8; Adenylate kinase isoenzyme 4, mitochondrial (ATP-AMP transphosphorylase); AK 8; AK8; ATP-AMP transphosphorylase 8; C9orf98; DDX31; HypB, AroK, ADK and rho factor domain containing protein RGD1303144; putative adenylate kinase-like protein C9orf98; putative adenylate kinase-like protein C9orf98 homolog; RGD1303144; RP11-143F18.1 |
| Gene Symbols | Ak8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.