Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adenylate Kinase 9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556563
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556563 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556563 Supplier Invitrogen™ Supplier No. PA556563
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VMAEHNPQYL IELNGNKPAE ELFMIVMDRL KYLNLKRAAI LTKLQGAEEE INDTMENDEL FRTLASYKLI APRYRWQRSK WG Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 72%.

Involved in maintaining the homeostasis of cellular nucleotides by catalyzing the interconversion of nucleoside phosphates. Has both nucleoside monophosphate and diphosphate kinase activities. Catalyzes the phosphorylation of AMP, dAMP, CMP and dCMP with ATP as phosphate donor and of CMP with GTP as phosphate donor. Also catalyzes the production of ATP, CTP, GTP, UTP, dATP, dCTP, dGTP and TTP from the corresponding diphosphate substrates with either ATP or GTP as phosphate donor. Shows substrate preference of CDP > UDP > ADP > GDP > TDP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adenylate Kinase 9
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AK9
Gene Accession No. Q5TCS8
Gene Alias Adenylate kinase 9; adenylate kinase domain containing 1; adenylate kinase domain containing 2; Adenylate kinase domain-containing protein 1; Adenylate kinase domain-containing protein 2; AK 9; AK9; AKD1; AKD2; C6orf199; C6orf224; dJ70A9.1; Gm234; Gm7127; RGD1560386; RP1-70A9.1; similar to novel protein
Gene Symbols AK9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Adenylate Kinase 9. Recombinant protein control fragment (Product #RP-94910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221264
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.