Learn More
Invitrogen™ Adenylate Kinase 9 Polyclonal Antibody

Description
Immunogen sequence: CIDKLCITPQ ELLSRLGEFE QFCPVSLAES QELFDCSATD SLEFAAEFRG HYYKMSSQEK LNKFLENPEL YVPPLAPHPL PSADMIPKRL TLSELKSRFP KCA Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Specifications
Specifications
| Antigen | Adenylate Kinase 9 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AK9 |
| Gene Accession No. | Q5TCS8 |
| Gene Alias | Adenylate kinase 9; adenylate kinase domain containing 1; adenylate kinase domain containing 2; Adenylate kinase domain-containing protein 1; Adenylate kinase domain-containing protein 2; AK 9; AK9; AKD1; AKD2; C6orf199; C6orf224; dJ70A9.1; Gm234; Gm7127; RGD1560386; RP1-70A9.1; similar to novel protein |
| Gene Symbols | AK9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.