Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylosuccinate Lyase Antibody (001), DyLight 550 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289877R
Description
Adenylosuccinate Lyase Monoclonal antibody specifically detects Adenylosuccinate Lyase in Human samples. It is validated for ELISASpecifications
Adenylosuccinate Lyase | |
Monoclonal | |
DyLight 550 | |
adenylosuccinase, adenylosuccinate lyase, AMPS, ASase, ASL, EC 4.3.2, EC 4.3.2.2 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Adenylosuccinate Lyase (P30566-1; Met1-Leu484). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
158 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
001 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction