Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylosuccinate Lyase Antibody (16C10), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259414H
This item is not returnable.
View return policy
Description
Adenylosuccinate Lyase Monoclonal antibody specifically detects Adenylosuccinate Lyase in Human samples. It is validated for Western Blot, ELISASpecifications
Adenylosuccinate Lyase | |
Monoclonal | |
HRP | |
adenylosuccinase, adenylosuccinate lyase, AMPS, ASase, ASL, EC 4.3.2, EC 4.3.2.2 | |
Recombinant human Adenylosuccinate Lyase (1-484aa) purified from E. coli | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
158 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
16C10 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction