Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylosuccinate Lyase Antibody (OTI2D10), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270101V
Description
Adenylosuccinate Lyase Monoclonal specifically detects Adenylosuccinate Lyase in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Adenylosuccinate Lyase | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADSL | |
Full length recombinant protein of human Adenylosuccinate Lyase (NP_000017) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI2D10 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
adenylosuccinase, adenylosuccinate lyase, AMPS, ASase, ASL, EC 4.3.2, EC 4.3.2.2 | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
158.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction