Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylosuccinate Lyase Antibody (OTI2D10), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270101MFV450
Description
Adenylosuccinate Lyase Monoclonal antibody specifically detects Adenylosuccinate Lyase in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow CytometrySpecifications
Adenylosuccinate Lyase | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
Purified |
Flow Cytometry | |
OTI2D10 | |
Flow Cytometry | |
adenylosuccinase, adenylosuccinate lyase, AMPS, ASase, ASL, EC 4.3.2, EC 4.3.2.2 | |
Full length human recombinant protein of human ADSL (NP_000017) produced in HEK293T cell. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
158 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction