Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Adenylosuccinate lyase Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
 
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | Adenylosuccinate lyase | 
|---|---|
| Concentration | 0.22 mg/mL | 
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | 
| Classification | Polyclonal | 
| Conjugate | Unconjugated | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
| 50-173-0102    | Proteintech Group Inc 152641AP150UL | 150 μL | 
                                        
                                            
                                                
                                                
                                                
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each of 1 for $518.60
                                                 |  | |||||
Description
ADSL (adenylosuccinate lyase), also known as AMPS, ASL or ASASE, is a 484 amino acid protein that is involved in both purine biosynthesis and in the formation of adenosine monophosphate (AMP) from inosine monophosphate. Expressed ubiquitously, ADSL catalyzes two key reactions in AMP biosynthesis, namely the removal of a fumarate from succinylaminoimidazole carboxamide (SAICA) ribotide to give aminoimidazole carboxamide ribotide (AICA) and the subsequent removal of fumarate from adenylosuccinate to yield AMP. Defects in the gene encoding ADSL are the cause of adenylosuccinase deficiency (ADSL deficiency), an autosomal recessive disorder characterized by epilepsy, growth retardation and muscular wasting. Multiple isoforms of ADSL exist due to alternative splicing events.Specifications
| Adenylosuccinate lyase | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P30566, P54822 | |
| 11564, 158, 315150 | |
| Adenylosuccinate lyase Fusion Protein Ag7332 | |
| Primary | |
| -20°C | |
| ADSL | 
| 0.22 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Adenylosuccinase, adenylosuccinate lyase, ADSL, AMPS, ASASE, ASL | |
| ADSL | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
			
             
                                                 
                                                 
                                                 
                                                 
                                                