Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adenylosuccinate Synthase Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310839100UL
Description
Adenylosuccinate Synthase Polyclonal specifically detects Adenylosuccinate Synthase in Human samples. It is validated for Western Blot.Specifications
Adenylosuccinate Synthase | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
ADEH, adenylosuccinate synthase, adenylosuccinate synthetase (Ade(-)H-complementing), adenylosuccinate synthetase isozyme 2, Adenylosuccinate synthetase, acidic isozyme, Adenylosuccinate synthetase, liver isozyme, AdSS 2, ADSS2, AMPSase 2, EC 6.3.4.4, IMP--aspartate ligase 2, L-type adenylosuccinate synthetase, MGC20404 | |
The immunogen is a synthetic peptide directed towards the middle region of Mouse Adenylosuccinate Synthase (NP_031448.2). Peptide sequence GWEKRLIISDRAHIVFDFHQAADGIQEQQRQEQAGKNLGTTKKGIGPVYS | |
100 μg | |
Lipid and Metabolism | |
159 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction