Learn More
NOW Diagnostics ADEXUSDx™ hCG Test

Description
- Uses whole blood (capillary or venous), plasma, or serum
- >99% sensitivity and specificity
- Clinical relevant cutoff: 10 mIU/mL
- Small sample volume test (about 40 μL)
- Results in 10 minutes, no waiting on urine samples
- No reagents or buffers
- Reduced exposure to liability from false negatives
- Increased patient throughput
Certifications
FDA Approved in USA; CE Marked in Europe
Specifications
Specifications
| Content And Storage | Stable unopened at 4°C-30°C (39°F-86°F) until the expiration date. |
| CE Marker | Yes |
| Clia Complexity | Moderate (Waiver Pending) |
| Detectable Analytes | hCG |
| Sample Type | Capillary or Venous blood/whole blood/plasma/serum |
| Sensitivity | 10 mIU/mL |
| Type | Rapid Diagnostic Lateral Flow Assay |
| Disposable | Single-use |
| Formulation | The device contains material of human or animal origin |
| For Use With (Application) | For in vitro diagnostic use only |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.