Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADFP Mouse anti-Human, Clone: ADFP/1366, Abnova™

Catalog No. 89034824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-824 Supplier Abnova Corporation Supplier No. MAB14741
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human ADFP.

  • High specificity
  • High reactivity

Specifications

Antigen ADFP
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone ADFP/1366
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human ADFP.
Dilution ELISA Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. Q99541
Gene Alias ADRP/MGC10598
Gene Symbols ADFP
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 249-376 of human ADFP.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.