Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADFP Rabbit anti-Human, Unconjugated, Clone: ADFP/2755R, Abnova™

Catalog No. 89337824 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-337-824 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-337-824 Supplier Abnova Corporation Supplier No. RAB00399
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

ADFP Monoclonal antibody specifically detects ADFP in Human samples. It is validated for WB, IHC-P.

Specifications

Antigen ADFP
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ADFP/2755R
Conjugate Unconjugated
Description adipose differentiation-related protein
Gene ADFP
Gene Alias ADRP, MGC10598
Host Species Rabbit
Immunogen Original antibody is raised against recombinant protein corresponding to amino acids 249-376 of human ADFP
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 123
Target Species Human
Content And Storage Store at 2 to 8°C
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.