Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADFP Recombinant Rabbit Monoclonal Antibody (HL2145)
GREENER_CHOICE

Catalog No. FEMA550400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550400 Supplier Invitrogen™ Supplier No. MA550400
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

ADFP Recombinant Monoclonal Antibody for Western Blot, IHC (P)

Milk lipid globules from humans, cows and rats contain a protein identified as adipocyte differentiation-related protein (ADFP). It is associated with the globule surface membrane material. This protein, previously believed to be specific to adipocytes, is a major constituent of the globule surface and is present in a detergent-insoluble complex that contains butyrophilin and xanthine oxidase. ADFP (Adipophilin) is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells. In tissues, however, expression of adipophilin is restricted to specific cell types, such as lactating mammary epithelial cells, adrenal cortex cells, Sertoli and Leydig cells of the male reproductive system, and steatosis or fatty change hepatocytes in alcoholic liver cirrhosis. ADFP may be a possible new marker for the identification of specialized differentiated cells containing lipid droplets and for diseases associated with fat-accumulating cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADFP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2145
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Plin2
Gene Accession No. P43883, Q99541
Gene Alias AA407157; Adfp; Adipophilin; adipose differentiation related protein; adipose differentiation-related protein; ADPH; Adrp; ADRP antibody; MGC10598; perilipin 2; perilipin-2; Plin2
Gene Symbols Plin2
Host Species Rabbit
Immunogen Recombinant fragment of human ADFP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11520, 123
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.