Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADH1A/ADH1B/ADH1C Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142126
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142126 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5142126 Supplier Invitrogen™ Supplier No. PA5142126
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise the alpha (ADH1A, NP_ 000658.1), the beta (ADH1B, NP_000659.2) and gamma (ADH1C, NP_000660.1) polypeptide variants of human alcohol dehydrogenase. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

Alcohol dehydrogenase,consisting of several homo- and heterodimers of alpha, beta, and gamma subunits, exhibits high activity for ethanol oxidation and plays a major role in ethanol catabolism.Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Three genes encoding alpha, beta and gamma subunits are tandemly organized in a genomic segment as a gene cluster.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADH1A/ADH1B/ADH1C
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ADH1B
Gene Accession No. P00325, P00326, P07327
Gene Alias ADH, alpha subunit; ADH, beta subunit; ADH, gamma subunit; ADH1; ADH1A; ADH1B; ADH1C; ADH2; ADH3; alcohol dehydrogenase 1 (class I), alpha polypeptide; alcohol dehydrogenase 1A; alcohol dehydrogenase 1A (class I), alpha polypeptide; alcohol dehydrogenase 1B; alcohol dehydrogenase 1B (class I), beta polypeptide; alcohol dehydrogenase 1C; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2 (class I), beta polypeptide; alcohol dehydrogenase 3 (class I), gamma polypeptide; alcohol dehydrogenase subunit alpha; Alcohol dehydrogenase subunit beta; Alcohol dehydrogenase subunit gamma; aldehyde reductase; All-trans-retinol dehydrogenase [NAD(+)] ADH1B; epididymis secretory protein Li 117; HEL-S-117
Gene Symbols ADH1A, ADH1B, ADH1C
Host Species Goat
Immunogen Peptide with sequence STAGKVMKCKA.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124, 125, 126
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.