Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542387
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542387 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542387 Supplier Invitrogen™ Supplier No. PA542387
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PLTNLCGKIS NLKSPASDQQ LMEDKTSRFT CKGKPVYHFF GTSTFSQYTV Sequence homology: Human: 100%.

This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADH4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ADH4
Gene Accession No. P08319
Gene Alias Ac1002; ADH-1; ADH2; ADH-2; Adh4; Alcohol dehydrogenase 4; alcohol dehydrogenase 4 (class II), pi polypeptide; alcohol dehydrogenase class II; Alcohol dehydrogenase class II pi chain; alcohol dehydrogenase II; aldehyde reductase; all-trans-retinol dehydrogenase [NAD(+)] ADH4; class II alcohol dehydrogenase, pi subunit; class II type ADH; epididymis secretory protein Li 4; HEL-S-4
Gene Symbols ADH4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ADH4.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.