Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADH4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596329
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596329 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596329 Supplier Invitrogen™ Supplier No. PA596329
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGTKGKVIKC KAAIAWEAGK PLCIEEVEVA PPKAHEVRIQ IIATSLCHTD ATVIDSKFEG LAFPVIVGHE AAGIVESIGP GVTNVKPGDK VIPLYAPLCR KCKFCLSPLT NLCGKISNLK SPASDQQLME DKTSRFTCKG KPVYHFFGTS TFSQYTVVSD INLAKIDDDA NLERVCLLGC GFSTGYGAAI NNAKVTPGST CAVFGLGGVG LSAVMGCKAA GASRIIGIDI NSEKFVKAKA LGATDCLNPR DLHKPIQEVI IELTKGGVDF ALDCAGGSET MKAALDCTTA GWGSCTFIGV AAGSKGLTIF PEELIIGRTI NGTFFGGWKS VDSIPKLVTD YKNKKFNLDA LVTHTLPFDK ISEAFDLMNQ GKSVRTILIF; Positive Samples: Mouse liver; Cellular Location: Cytoplasm.

This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADH4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADH4
Gene Accession No. Q9QYY9
Gene Alias Ac1002; ADH-1; ADH2; ADH-2; Adh4; Alcohol dehydrogenase 4; alcohol dehydrogenase 4 (class II), pi polypeptide; alcohol dehydrogenase class II; Alcohol dehydrogenase class II pi chain; alcohol dehydrogenase II; aldehyde reductase; all-trans-retinol dehydrogenase [NAD(+)] ADH4; class II alcohol dehydrogenase, pi subunit; class II type ADH; epididymis secretory protein Li 4; HEL-S-4
Gene Symbols ADH4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-380 of human ADH4 (NP_0006612).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26876
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.