Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADH5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597436
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597436 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597436 Supplier Invitrogen™ Supplier No. PA597436
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ADH5 Polyclonal Antibody for Western Blot, ELISA

This gene encodes a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The encoded protein forms a homodimer. It has virtually no activity for ethanol oxidation, but exhibits high activity for oxidation of long-chain primary alcohols and for oxidation of S-hydroxymethyl-glutathione, a spontaneous adduct between formaldehyde and glutathione. This enzyme is an important component of cellular metabolism for the elimination of formaldehyde, a potent irritant and sensitizing agent that causes lacrymation, rhinitis, pharyngitis, and contact dermatitis. The human genome contains several non-transcribed pseudogenes related to this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADH5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ADH5
Gene Accession No. P28474
Gene Alias ADH1C; Adh2; Adh-2; ADH3; ADH-3; ADH5; Adh-5; ADH-B2; ADHX; alcohol dehydrogenase (class III), chi polypeptide; alcohol dehydrogenase 1C (class I), gamma polypeptide; alcohol dehydrogenase 2; Alcohol dehydrogenase 5; alcohol dehydrogenase 5 (class III), chi polypeptide; alcohol dehydrogenase B2; Alcohol dehydrogenase class chi chain; alcohol dehydrogenase class-3; Alcohol dehydrogenase class-III; class III alcohol dehydrogenase; class III alcohol dehydrogenase, chi subunit; epididymis secretory sperm binding protein Li 60p; FALDH; FDH; formaldehyde dehydrogenase; glutathione-dependent formaldehyde dehydrogenase; GSH-FDH; GSNOR; HEL-S-60p; S-(hydroxymethyl)glutathione dehydrogenase; S-nitrosoglutathione reductase
Gene Symbols ADH5
Host Species Rabbit
Immunogen Recombinant Mouse Alcohol dehydrogenase class-3 protein (2-374AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11532
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.