Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADH6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584801 Supplier Invitrogen™ Supplier No. PA584801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VADYMAEKLN LDPLITHTLN LDKINEAVEL MKTGKW Highest antigen sequence indentity to the following orthologs: Mouse - 61%, Rat - 67%.

This gene encodes class V alcohol dehydrogenase, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. This gene is expressed in the stomach as well as in the liver, and it contains a glucocorticoid response element upstream of its 5' UTR, which is a steroid hormone receptor binding site. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADH6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADH6
Gene Accession No. P28332
Gene Alias 1810009C16Rik; ADH-5; Adh5a; ADH6; Adh6a; Alcohol dehydrogenase 6; alcohol dehydrogenase 6 (class V); alcohol dehydrogenase 6A (class V); aldehyde reductase; class V alcohol dehydrogenase
Gene Symbols ADH6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADH6. Recombinant protein control fragment (Product #RP-108244).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.