Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADH7 Monoclonal Antibody (OTI2D11), Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Thermo Scientific MA526001

Catalog No. PIMA526001


Only null left
Add to Cart

Description

Description

This gene encodes class IV alcohol dehydrogenase 7 mu or sigma subunit, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The enzyme encoded by this gene is inefficient in ethanol oxidation, but is the most active as a retinol dehydrogenase; thus it may participate in the synthesis of retinoic acid, a hormone important for cellular differentiation. The expression of this gene is much more abundant in stomach than liver, thus differing from the other known gene family members.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ADH7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P40394
ADH7
Full length human recombinant protein of ADH7 produced in HEK293T cell
100μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI2D11
Unconjugated
ADH7
Adh3; Adh-3; Adh-3e; Adh3-e; Adh-3t; Adh3-t; Adh4; Adh7; ADH-C2; Adt-1; AI325182; alcohol dehydrogenase 3 complex; alcohol dehydrogenase 3 temporal; alcohol dehydrogenase 4; Alcohol dehydrogenase 7; alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide; alcohol dehydrogenase class 4 mu/sigma chain; Alcohol dehydrogenase class IV mu/sigma chain; alcohol dehydrogenase VII; alcohol dehydrogenase-7; All-trans-retinol dehydrogenase [NAD(+)] ADH7; class IV alcohol dehydrogenase, mu or sigma subunit; class IV sigma-1 alcohol dehydrogenase; class IV sigmasigma alcohol dehydrogenase; gastric alcohol dehydrogenase; IV ADH; Omega-hydroxydecanoate dehydrogenase ADH7; ret; Retinol dehydrogenase
Mouse
Affinity Chromatography
RUO
131
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.