Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADH7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558887
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558887 Supplier Invitrogen™ Supplier No. PA558887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFEKAMAVGA TECISPKDST KPISEVLSEM TGNNVGYTFE VIGHLETMID ALASCHMNYG TSV Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 81%.

This gene encodes class IV alcohol dehydrogenase 7 mu or sigma subunit, which is a member of the alcohol dehydrogenase family. Members of this family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. The enzyme encoded by this gene is inefficient in ethanol oxidation, but is the most active as a retinol dehydrogenase; thus it may participate in the synthesis of retinoic acid, a hormone important for cellular differentiation. The expression of this gene is much more abundant in stomach than liver, thus differing from the other known gene family members.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADH7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ADH7
Gene Accession No. P40394, P41682, Q64437
Gene Alias Adh3; Adh-3; Adh-3e; Adh3-e; Adh-3t; Adh3-t; Adh4; Adh7; ADH-C2; Adt-1; AI325182; alcohol dehydrogenase 3 complex; alcohol dehydrogenase 3 temporal; alcohol dehydrogenase 4; Alcohol dehydrogenase 7; alcohol dehydrogenase 7 (class IV), mu or sigma polypeptide; alcohol dehydrogenase class 4 mu/sigma chain; Alcohol dehydrogenase class IV mu/sigma chain; alcohol dehydrogenase VII; alcohol dehydrogenase-7; All-trans-retinol dehydrogenase [NAD(+)] ADH7; class IV alcohol dehydrogenase, mu or sigma subunit; class IV sigma-1 alcohol dehydrogenase; class IV sigmasigma alcohol dehydrogenase; gastric alcohol dehydrogenase; IV ADH; Omega-hydroxydecanoate dehydrogenase ADH7; ret; Retinol dehydrogenase
Gene Symbols ADH7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ADH7. Recombinant protein control fragment (Product #RP-96754).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11529, 131, 171178
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.