Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Adhesive Seals

Catalog No. 14387312
Change view
Click to view available options
Material:
Polyethylene
For Use With (Application):
Crystallography and Genotyping
Fluorescence Applications, QPCR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material For Use With (Application)
14-387-312 Polyethylene Fluorescence Applications, QPCR
14-387-313 Polyethylene Crystallography and Genotyping
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-387-312 Supplier Thermo Scientific™ Supplier No. AB2170
Add to Cart
Add to Cart
Request a Quote

Perform high throughput applications at a lower cost per plate with these adhesive seals, featuring an automated format provides efficient sealing.

Perform high throughput applications at a lower cost per plate with Thermo Scientific™ Adhesive Seals. An automated format provides efficient sealing. Rolls are designed for use with the Thermo Scientific™ SEAL-IT 100™ adhesive sealer.

Adhesive PCR Film Roll
  • Clear adhesive seal prevents evaporation during sample storage or thermal cycling, while still allowing samples to be accessed by piercing foil with pipette tips
  • No adhesive build-up on tips after piercing
  • PCR Film seals are recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -80° to 120°C

Adhesive PCR Foil Roll

  • Strong adhesive seal prevents evaporation during sample storage or thermal cycling, while allowing sample access by piercing foil with pipet tips
  • No adhesive build-up on tips after piercing
  • PCR Foil Seals are recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -80° to +150°C

Adhesive PCR Film

  • Optically clear adhesive plate seal for sample storage; thermal cycling with screw down or clip down lid
  • Strong adhesive, peelable with no sticky residue left on the plate
  • Temperature range: -20°C to +120°C

ABsolute QPCR Seal Roll

  • Optically clear adhesive plate seal is designed for fluorescence applications, including QPCR
  • Features strong pressure-sensitive, non-sticky adhesive that prevents evaporation during sample storage or thermal cycling
  • Seals are easily peelable, but leave no sticky residue on the plate when removed
  • Recommended for use in conjunction with a cycler that has a screw-down or clip-down lid
  • Temperature range: -80° to +110°C

Adhesive Crystallography Seal Roll

  • Optically clear, pressure sensitive, non-sticky adhesive seal for crystallography and genotyping applications
  • Samples are not exposed to adhesive
  • Seal is easily peelable without leaving residue on the plate
  • Temperature range: -70° to +110C°
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Description ABsolute QPCR Seal Roll
Material Polyethylene
For Use With (Application) Fluorescence Applications, QPCR
For Use With (Equipment) The Seal-it 100 adhesive sealer.
Product Type ABsolute QPCR Seal
Pierceable Yes
Temperature Range (Metric) -80°C to +110°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.