Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADHFE1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554682
Description
Immunogen sequence: LEMAEILGAD TRTARIQDAG LVLADTLRKF LFDLDVDDGL AAVGYSKADI PALVKGTLPQ ERVTKLAPCP QSEEDLAALF EASMKL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
The ADHFE1 gene encodes hydroxyacid-oxoacid transhydrogenase (EC 1.1.99.24), which is responsible for the oxidation of 4-hydroxybutyrate in mammalian tissues (Kardon et al., 2006 ).
Specifications
ADHFE1 | |
Polyclonal | |
Unconjugated | |
Adhfe1 | |
6330565B14Rik; ADH8; ADHFe1; AI043035; alcohol dehydrogenase 8; alcohol dehydrogenase iron-containing protein 1; alcohol dehydrogenase, iron containing 1; alcohol dehydrogenase, iron containing, 1; Fe-containing alcohol dehydrogenase; Fe-containing alcohol dehydrogenase 1; HMFT2263; HOT; hydroxyacid-oxoacid transhydrogenase, mitochondrial | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
137872, 362474, 76187 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4QQW3, Q8IWW8, Q8R0N6 | |
Adhfe1 | |
Recombinant protein corresponding to Human ADHFE1. Recombinant protein control fragment (Product #RP-93812). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction