Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADI1 Antibody (27E8), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP222614AF594

Catalog No. NP222614A94


Only null left
Add to Cart

Description

Description

ADI1 Monoclonal specifically detects ADI1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ADI1
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
ADI1
Recombinant human ADI1 (1-179aa) purified from E. coli
metabolism, Signal Transduction
55256
Store at 4°C in the dark.
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
2700000000
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase, Acireductone dioxygenase (Ni(2+)-requiring), acireductone dioxygenase 1, APL1, ARDsubmergence induced protein 2, EC 1.13.11.53, FLJ10913, HMFT1638, membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1, Membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1, MTCBP1, MTCBP-1, Ni-ARD, SIPLMT1-MMP cytoplasmic tail-binding protein-1, Submergence-induced protein 2 homolog
Mouse
0.1 mL
Primary
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.