Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADI1 Antibody (OTI5C2), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271994G
Description
ADI1 Monoclonal specifically detects ADI1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
ADI1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ADI1 | |
Full length recombinant protein of human ADI1 (NP_060739) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI5C2 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase, Acireductone dioxygenase (Ni(2+)-requiring), acireductone dioxygenase 1, APL1, ARDsubmergence induced protein 2, EC 1.13.11.53, FLJ10913, HMFT1638, membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1, Membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1, MTCBP1, MTCBP-1, Ni-ARD, SIPLMT1-MMP cytoplasmic tail-binding protein-1, Submergence-induced protein 2 homolog | |
Mouse | |
Affinity Purified | |
metabolism, Signal Transduction | |
55256.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction