Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ADI1 Monoclonal Antibody (OTI5C2)
GREENER_CHOICE

Catalog No. PIMA525269
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525269 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525269 Supplier Invitrogen™ Supplier No. MA525269
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ADI1 Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene encodes an enzyme that belongs to the aci-reductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADI1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5C2
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ADI1
Gene Accession No. Q9BV57
Gene Alias 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase; 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase {ECO:0000255; aci-reductone dioxygenase; aci-reductone dioxygenase (ARD)-like protein 1; acireductone dioxygenase (Fe(2+)-requiring); acireductone dioxygenase (Fe(2+)-requiring) {ECO:0000255; acireductone dioxygenase (Ni(2+)-requiring); acireductone dioxygenase 1; ADI1; AL024210; Alp1; Androgen-responsive ARD-like protein 1; androgen-responsive gene encoding an ARD-like protein; APL1; ARD; ARD {ECO:0000255; fe-ARD; fe-ARD {ECO:0000255; HAMAP-Rule:MF_03154}; HMFT1638; membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1 {ECO:0000255; MT1-MMP cytoplasmic tail-binding protein-1; MTCBP1; MTCBP-1; MTCBP-1 {ECO:0000255; mtnD; Ni-ARD; SIPL; Sip-L; submergence induced protein 2; submergence-induced protein-like factor
Gene Symbols ADI1
Host Species Mouse
Immunogen Full length human recombinant protein of ADI1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55256
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.