Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ADI1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531217
Description
Recommended positive controls: HepG2, mouse lung, rat lung. Predicted reactivity: Mouse (88%), Rat (87%), Pig (88%), Rabbit (88%), Chicken (83%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an enzyme that belongs to the aci-reductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.
Specifications
ADI1 | |
Polyclonal | |
Unconjugated | |
ADI1 | |
1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase; 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase {ECO:0000255; aci-reductone dioxygenase; aci-reductone dioxygenase (ARD)-like protein 1; acireductone dioxygenase (Fe(2+)-requiring); acireductone dioxygenase (Fe(2+)-requiring) {ECO:0000255; acireductone dioxygenase (Ni(2+)-requiring); acireductone dioxygenase 1; ADI1; AL024210; Alp1; Androgen-responsive ARD-like protein 1; androgen-responsive gene encoding an ARD-like protein; APL1; ARD; ARD {ECO:0000255; fe-ARD; fe-ARD {ECO:0000255; HAMAP-Rule:MF_03154}; HMFT1638; membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1 {ECO:0000255; MT1-MMP cytoplasmic tail-binding protein-1; MTCBP1; MTCBP-1; MTCBP-1 {ECO:0000255; mtnD; Ni-ARD; SIPL; Sip-L; submergence induced protein 2; submergence-induced protein-like factor | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
104923, 298934, 55256 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q562C9, Q99JT9, Q9BV57 | |
ADI1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 179 of Human ADI1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction