Learn More
Invitrogen™ ADI1 Polyclonal Antibody

Description
Immunogen sequence: FYEEHLHLDD EIRYILDGSG YFDVRDKEDQ WIRIFMEKGD MVTLPAGIYH RFTVDEKNYT KAMRLFVGEP VWTAYNRPAD HFEAR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Specifications
Specifications
| Antigen | ADI1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ADI1 |
| Gene Accession No. | Q9BV57 |
| Gene Alias | 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase; 1,2-dihydroxy-3-keto-5-methylthiopentene dioxygenase {ECO:0000255; aci-reductone dioxygenase; aci-reductone dioxygenase (ARD)-like protein 1; acireductone dioxygenase (Fe(2+)-requiring); acireductone dioxygenase (Fe(2+)-requiring) {ECO:0000255; acireductone dioxygenase (Ni(2+)-requiring); acireductone dioxygenase 1; ADI1; AL024210; Alp1; Androgen-responsive ARD-like protein 1; androgen-responsive gene encoding an ARD-like protein; APL1; ARD; ARD {ECO:0000255; fe-ARD; fe-ARD {ECO:0000255; HAMAP-Rule:MF_03154}; HMFT1638; membrane-type 1 matrix metalloproteinase cytoplasmic tail binding protein-1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1; membrane-type 1 matrix metalloproteinase cytoplasmic tail-binding protein 1 {ECO:0000255; MT1-MMP cytoplasmic tail-binding protein-1; MTCBP1; MTCBP-1; MTCBP-1 {ECO:0000255; mtnD; Ni-ARD; SIPL; Sip-L; submergence induced protein 2; submergence-induced protein-like factor |
| Gene Symbols | ADI1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.