Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 151331AP150UL
Description
This gene encodes an enzyme that belongs to the acireductone dioxygenase family of metal-binding enzymes, which are involved in methionine salvage. This enzyme may regulate mRNA processing in the nucleus, and may carry out different functions depending on its localization. A pseudogene that is located on chromosome 20 has been defined for this gene.Specifications
| ADI1 | |
| Polyclonal | |
| Unconjugated | |
| ADI1 | |
| Acireductone dioxygenase, acireductone dioxygenase 1, ADI1, APL1, ARD, HMFT1638, MTCBP 1, MTCBP1, SIP L, SIPL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104923, 298934, 55256 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q562C9, Q99JT9, Q9BV57 | |
| ADI1 | |
| ADI1 Fusion Protein Ag7274 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction