Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ADIG Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP275311
Description
ADIG Polyclonal specifically detects ADIG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
ADIG | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
adipogenesis associated, adipogenin, KIAA1219, MGC149650, MGC39724, RP5-1100H13.2, SMAF1, small adipocyte factor 1 (SMAF1) | |
Goat | |
Affinity Purified | |
Primary | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/ml | |
Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA 1:2000 | |
Q0VDE8, NM_001018082 | |
ADIG | |
Peptide with sequence SQESKEDDSDLC, from the internal region of the protein sequence according to NP_663610.1. | |
0.1 mg | |
149685.0 | |
Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction