Learn More
Invitrogen™ Adipolean (gAcrp30) Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Goat Polyclonal Antibody
Supplier: Invitrogen™ 500P193GBT500UG
Description
AA Sequence of recombinant protein: MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human gAcrp30/Adipolean. Anti-Human gAcrp30/Adipolean-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hAdipolean by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human Adipolean (500-P193G) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hAdipolean. Western Blot: To detect Human Adipolean by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Adipolean is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P193GBT-1MG will be provided as 2 x 500 μg
gAcrp30 is a naturally occurring globular protein obtained by proteolytic processing of adiponectin. Adiponectin is produced and secreted exclusively by adipocytes, and is a relatively abundant plasma protein, accounting for up to 0.05% of total serum protein. Like Adiponectin, gAcrp30 is capable of decreasing hyperglycemia and reversing insulin resistance. Additionally, gAcrp30 has been shown to be an important factor in promoting fat loss by signaling muscle to absorb and burn Free-Fatty Acids (FFAs). The signaling receptors for adiponectin and gAcrp30 have recently been identified and named AdipoR1 and AdipoR2. AdipoR2 is predominantly expressed in the liver.
Specifications
| Adipolean (gAcrp30) | |
| Polyclonal | |
| Biotin | |
| ADIPOQ | |
| gAcrp30 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 9370 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| Q15848 | |
| ADIPOQ | |
| E.coli-derived Recombinant Human gAcrp30/Adipolean. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.