Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Adiponectin/Acrp30 Antibody (ADPN/1370), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | Adiponectin/Acrp30 |
---|---|
Clone | ADPN/1370 |
Dilution | Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page, Immunofluorescence 1-2ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP247652F
![]() |
Novus Biologicals
NBP2476520.02MG |
0.02 mg |
Each for $268.00
|
|
|||||
NBP247652A
![]() |
Novus Biologicals
NBP2476520.1MG |
0.1 mg |
Each for $524.00
|
|
|||||
NBP247652B
![]() |
Novus Biologicals
NBP2476520.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Adiponectin/Acrp30 Monoclonal specifically detects Adiponectin/Acrp30 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
Adiponectin/Acrp30 | |
Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page, Immunofluorescence 1-2ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACDC, ACRP30ADPN, adipocyte, C1Q and collagen domain containing, adiponectin, adiponectin, C1Q and collagen domain containing, AdipoQ, ADIPQTL1, apM-1, APM1APM-1, C1q and collagen domain-containing protein, GBP28apM1, Gelatin-binding protein | |
ADIPOQ | |
IgG2b | |
Protein A or G purified | |
This MAb reacts with adiponectin, an adipocytokine. Adipocytokines are hormones produced in adipose tissue. Adiponectin is abundantly present in plasma and has insulin like effect on glucose levels in the blood. Plasma adiponectin levels are low in insulin resistant patients who are obese, have diabetes mellitus type 2 or HIV-lipodystrophy. In women adiponectin levels tend to be higher than in men, which may be due to androgens suppressing adiponectin levels. Furthermore adiponectin and leptin are both indicated in regulating body weight through direct action on the hypothalamus, influencing appetite. Obese people have low adiponectin levels while levels in anorexia patients are high. Adiponectin acts as ligand for various receptors, two of which have been identified, one probably involved in carbohydrate assimilation, the other in tuning the rate of metabolism. |
ADPN/1370 | |
Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
Unconjugated | |
Mouse | |
Diabetes Research, Lipid and Metabolism | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
9370 | |
Recombinant human Adiponectin protein fragment | |
Primary | |
Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title